siRNA Details
| Gene Symbol: Spcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131006.1 | NP_001124478.1 | 4 | 500 | ||
| GenBank mRNA | CF976587.1 | - | 4 | 506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spcs1| Gene Symbol: Spcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131006.1 | NP_001124478.1 | 4 | 500 | ||
| GenBank mRNA | CF976587.1 | - | 4 | 506 | ||