siRNA Details
| Gene Symbol: Arvcf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131013.1 | NP_001124485.1 | 18 | 3699 | ||
| XM_008768880.1 | XP_008767102.1 | 19 | 3708 | |||
| XM_008768881.1 | XP_008767103.1 | 18 | 4672 | |||
| XM_008768882.1 | XP_008767104.1 | 17 | 3818 | |||
| XM_008768885.1 | XP_008767107.1 | 17 | 5949 | |||
| GenBank mRNA | BC168253.1 | - | 18 | 3699 | 5621230 | |

Arvcf
Genomic Map