siRNA Details
| Gene Symbol: Tpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134994.1 | NP_001128466.1 | 5 | 395 | ||
| XM_008762921.1 | XP_008761143.1 | 2 | 1941 | |||
| GenBank mRNA | BC168695.1 | - | 5 | 284 | 7938123 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpk1| Gene Symbol: Tpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134994.1 | NP_001128466.1 | 5 | 395 | ||
| XM_008762921.1 | XP_008761143.1 | 2 | 1941 | |||
| GenBank mRNA | BC168695.1 | - | 5 | 284 | 7938123 | |