siRNA Details
| Gene Symbol: Zfp951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134998.1 | NP_001128470.1 | 1663 | |||
| GenBank mRNA | BC169037.1 | - | 1663 | 7368372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp951| Gene Symbol: Zfp951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134998.1 | NP_001128470.1 | 1663 | |||
| GenBank mRNA | BC169037.1 | - | 1663 | 7368372 | ||