siRNA Details
Gene Symbol: Zfp951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134998.1 | NP_001128470.1 | 472 | |||
GenBank mRNA | BC169037.1 | - | 472 | 7368372 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134998.1 | NP_001128470.1 | 472 | |||
GenBank mRNA | BC169037.1 | - | 472 | 7368372 |