siRNA Details
| Gene Symbol: Flna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134599.1 | NP_001128071.1 | 41, 42 | 7177 | ||
| XM_006229569.2 | XP_006229631.1 | 43, 44 | 7255 | |||
| XM_006229570.2 | XP_006229632.1 | 42, 43 | 7231 | |||
| GenBank mRNA | FJ416060.1 | - | 41, 42 | 7124 | ||
| FQ230835.1 | - | 18, 19 | 2785 | |||

Flna
Genomic Map