siRNA Details
| Gene Symbol: Flna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134599.1 | NP_001128071.1 | 33 | 5721 | ||
| XM_006229569.2 | XP_006229631.1 | 35 | 5799 | |||
| XM_006229570.2 | XP_006229632.1 | 34 | 5775 | |||
| GenBank mRNA | FJ416060.1 | - | 33 | 5668 | ||
| FQ230835.1 | - | 10 | 1329 | |||

Flna
Genomic Map