siRNA Details
| Gene Symbol: Dnajc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134640.1 | NP_001128112.1 | 5 | 393 | ||
| XM_006232259.2 | XP_006232321.1 | 5 | 408 | |||
| GenBank mRNA | FQ212146.1 | - | 5 | 352 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc19| Gene Symbol: Dnajc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134640.1 | NP_001128112.1 | 5 | 393 | ||
| XM_006232259.2 | XP_006232321.1 | 5 | 408 | |||
| GenBank mRNA | FQ212146.1 | - | 5 | 352 | ||