siRNA Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134777.1 | NP_001128249.1 | 9 | 1324 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pex19| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134777.1 | NP_001128249.1 | 9 | 1324 | ||