siRNA Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107375.2 | NP_001100845.1 | 8 | 2239 | ||
| Gene Symbol: LOC100911356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751804.3 | XP_003751852.1 | 8 | 2232 | ||
| XM_008772208.1 | XP_008770430.1 | 8 | 2252 | |||

Pex19
Genomic Map