siRNA Details
| Gene Symbol: Ormdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134704.1 | NP_001128176.1 | 672 | |||
| GenBank mRNA | BC166795.1 | - | 672 | 7937873 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ormdl1| Gene Symbol: Ormdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134704.1 | NP_001128176.1 | 672 | |||
| GenBank mRNA | BC166795.1 | - | 672 | 7937873 | ||