siRNA Details
Gene Symbol: Fam227a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765683.1 | XP_008763905.1 | 10 | 1360 | ||
GenBank mRNA | BC097470.1 | - | 11 | 1161 | 7457750 | |
CB546465.1 | - | 3 | 227 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam227a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765683.1 | XP_008763905.1 | 10 | 1360 | ||
GenBank mRNA | BC097470.1 | - | 11 | 1161 | 7457750 | |
CB546465.1 | - | 3 | 227 |