siRNA Details
| Gene Symbol: LOC685544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109474.1 | NP_001102944.1 | 1 | 599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC685544| Gene Symbol: LOC685544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109474.1 | NP_001102944.1 | 1 | 599 | ||