siRNA Details
| Gene Symbol: Ost4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134690.1 | NP_001128162.1 | 2 | 429 | ||
| NM_001134691.1 | NP_001128163.1 | 3 | 329 | |||
| GenBank mRNA | BC157814.1 | - | 2 | 407 | 8362186 | |
| CB706964.1 | - | 3 | 290 | |||

Ost4
Genomic Map