siRNA Details
| Gene Symbol: Vpreb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108845.1 | NP_001102315.1 | 2 | 387 | ||
| Gene Symbol: Vpreb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134788.1 | NP_001128260.1 | 2 | 263 | ||

Vpreb1
Genomic Map