siRNA Details
| Gene Symbol: Tgif2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134983.1 | NP_001128455.1 | 3 | 1625 | ||
| XM_006235424.2 | XP_006235486.1 | 2 | 1442 | |||
| XM_006235425.2 | XP_006235487.1 | 3 | 3514 | |||
| XM_006235426.2 | XP_006235488.1 | 3 | 1689 | |||
| XM_006235427.2 | XP_006235489.1 | 4 | 1594 | |||
| GenBank mRNA | BC168719.1 | - | 3 | 1625 | 8364226 | |
| FQ235168.1 | - | 3 | 1563 | |||

Tgif2
Genomic Map