siRNA Details
| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130580.1 | NP_001124052.1 | 5 | 1049 | ||
| XM_006241615.2 | XP_006241677.1 | 5 | 1015 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eny2| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130580.1 | NP_001124052.1 | 5 | 1049 | ||
| XM_006241615.2 | XP_006241677.1 | 5 | 1015 | |||