siRNA Details
Gene Symbol: LOC100174910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131003.1 | NP_001124475.1 | 4 | 706 | ||
XM_006254736.2 | XP_006254798.1 | 4 | 844 | |||
XM_006254737.2 | XP_006254799.1 | 3 | 787 | |||
XM_006254738.2 | XP_006254800.1 | 6 | 863 | |||
XM_006254739.2 | XP_006254801.1 | 6 | 856 | |||
XM_006254740.2 | XP_006254802.1 | 5 | 752 | |||
XM_006254741.2 | XP_006254803.1 | 5 | 749 | |||
XM_008772078.1 | XP_008770300.1 | 5 | 2219 | |||
XM_008772079.1 | XP_008770301.1 | 4 | 2118 | |||
GenBank mRNA | BC098717.1 | - | 3 | 1280 | ||
BC166916.1 | - | 3 | 649 | 6918206 | ||
BC168769.1 | - | 3 | 649 | 6891274 |
