siRNA Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134728.1 | NP_001128200.1 | 1 | 182 | ||
| GenBank mRNA | BC158602.1 | - | 1 | 182 | 7133303 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag16| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134728.1 | NP_001128200.1 | 1 | 182 | ||
| GenBank mRNA | BC158602.1 | - | 1 | 182 | 7133303 | |