siRNA Details
Gene Symbol: Hps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135612.1 | NP_001129084.1 | 23 | 3974 | ||
GenBank mRNA | BC086395.1 | - | 3 | 728 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135612.1 | NP_001129084.1 | 23 | 3974 | ||
GenBank mRNA | BC086395.1 | - | 3 | 728 |