siRNA Details
| Gene Symbol: LOC100125368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139487.1 | NP_001132959.1 | 4 | 363 | ||
| XM_006240835.2 | XP_006240897.1 | 4 | 537 | |||
| XM_008765080.1 | XP_008763302.1 | 4 | 796 | |||
| GenBank mRNA | BC128765.1 | - | 4 | 363 | 7123097 | |
| Gene Symbol: LOC100360514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765133.1 | XP_008763355.1 | 3 | 240 | ||
| XM_008765134.1 | XP_008763356.1 | 4 | 385 | |||
| Gene Symbol: LOC102551539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765145.1 | XP_008763367.1 | 261 | |||
| XM_008765146.1 | XP_008763368.1 | 796 | ||||

LOC100125368
Genomic Map