siRNA Details
| Gene Symbol: Zfp710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134563.1 | NP_001128035.1 | 1 | 552 | ||
| XM_008759507.1 | XP_008757729.1 | 2 | 2140 | |||
| XM_008759508.1 | XP_008757730.1 | 2 | 1945 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp710| Gene Symbol: Zfp710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134563.1 | NP_001128035.1 | 1 | 552 | ||
| XM_008759507.1 | XP_008757729.1 | 2 | 2140 | |||
| XM_008759508.1 | XP_008757730.1 | 2 | 1945 | |||