siRNA Details
| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134962.1 | NP_001128434.1 | 1 | 216 | ||
| XM_008759532.1 | XP_008757754.1 | 1 | 216 | |||
| GenBank mRNA | BC169015.1 | - | 1 | 216 | 7132705 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttll13| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134962.1 | NP_001128434.1 | 1 | 216 | ||
| XM_008759532.1 | XP_008757754.1 | 1 | 216 | |||
| GenBank mRNA | BC169015.1 | - | 1 | 216 | 7132705 | |