siRNA Details
| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134962.1 | NP_001128434.1 | 15 | 3341 | ||
| XM_008759532.1 | XP_008757754.1 | 15 | 3341 | |||
| XM_008759533.1 | XP_008757755.1 | 14 | 2949 | |||
| XM_008759534.1 | XP_008757756.1 | 15 | 2714 | |||
| XM_008759535.1 | XP_008757757.1 | 13 | 2681 | |||
| GenBank mRNA | BC169015.1 | - | 15 | 3341 | 7132705 | |

Ttll13
Genomic Map