siRNA Details
| Gene Symbol: Nrbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135007.1 | NP_001128479.1 | 9 | 1224 | ||
| XM_006241885.2 | XP_006241947.1 | 10 | 1243 | |||
| GenBank mRNA | FQ223130.1 | - | 4 | 785 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrbp2| Gene Symbol: Nrbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135007.1 | NP_001128479.1 | 9 | 1224 | ||
| XM_006241885.2 | XP_006241947.1 | 10 | 1243 | |||
| GenBank mRNA | FQ223130.1 | - | 4 | 785 | ||