siRNA Details
| Gene Symbol: LOC680200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139491.1 | NP_001132963.1 | 4 | 1551 | ||
| GenBank mRNA | BC167062.1 | - | 4 | 1551 | 9024521 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC680200| Gene Symbol: LOC680200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139491.1 | NP_001132963.1 | 4 | 1551 | ||
| GenBank mRNA | BC167062.1 | - | 4 | 1551 | 9024521 | |