siRNA Details
| Gene Symbol: Gng13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135918.1 | NP_001129390.1 | 2 | 153 | ||
| XM_006246034.2 | XP_006246096.1 | 2 | 297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gng13| Gene Symbol: Gng13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135918.1 | NP_001129390.1 | 2 | 153 | ||
| XM_006246034.2 | XP_006246096.1 | 2 | 297 | |||