siRNA Details
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053350.2 | NP_445802.2 | 15 | 1681 | ||
GenBank mRNA | AF121052.1 | - | 15 | 1586 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053350.2 | NP_445802.2 | 15 | 1681 | ||
GenBank mRNA | AF121052.1 | - | 15 | 1586 |