siRNA Details
| Gene Symbol: Ptges3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130989.1 | NP_001124461.1 | 8 | 1001 | ||
| XM_008765026.1 | XP_008763248.1 | 8 | 1046 | |||
| GenBank mRNA | BC166579.1 | - | 8 | 1001 | 7379926 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptges3| Gene Symbol: Ptges3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130989.1 | NP_001124461.1 | 8 | 1001 | ||
| XM_008765026.1 | XP_008763248.1 | 8 | 1046 | |||
| GenBank mRNA | BC166579.1 | - | 8 | 1001 | 7379926 | |