siRNA Details
Gene Symbol: Pdxp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135819.1 | NP_001129291.1 | 2 | 1381 | ||
GenBank mRNA | BC166563.1 | - | 2 | 1381 | 7316162 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdxp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135819.1 | NP_001129291.1 | 2 | 1381 | ||
GenBank mRNA | BC166563.1 | - | 2 | 1381 | 7316162 |