siRNA Details
| Gene Symbol: Wdr25l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135894.1 | NP_001129366.1 | 7 | 1727 | ||
| XM_006240544.2 | XP_006240606.1 | 8 | 1944 | |||
| XM_006240545.2 | XP_006240607.1 | 8 | 1789 | |||
| XM_006240546.2 | XP_006240608.1 | 8 | 1807 | |||
| XM_006240547.2 | XP_006240609.1 | 8 | 2015 | |||
| GenBank mRNA | BC166712.1 | - | 7 | 1727 | 7930656 | |

Wdr25l
Genomic Map