siRNA Details
| Gene Symbol: Wdr25l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135894.1 | NP_001129366.1 | 3 | 1047 | ||
| XM_006240544.2 | XP_006240606.1 | 4 | 1264 | |||
| XM_006240545.2 | XP_006240607.1 | 4 | 1109 | |||
| XM_006240546.2 | XP_006240608.1 | 4 | 1127 | |||
| XM_006240547.2 | XP_006240609.1 | 4 | 1335 | |||
| GenBank mRNA | BC166712.1 | - | 3 | 1047 | 7930656 | |

Wdr25l
Genomic Map