siRNA Details
| Gene Symbol: LOC688553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135915.1 | NP_001129387.1 | 7 | 1378 | ||
| GenBank mRNA | BC127536.1 | - | 7 | 1378 | 7134154 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC688553| Gene Symbol: LOC688553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135915.1 | NP_001129387.1 | 7 | 1378 | ||
| GenBank mRNA | BC127536.1 | - | 7 | 1378 | 7134154 | |