siRNA Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134695.1 | NP_001128167.1 | 3 | 211 | ||
| XM_008772971.1 | XP_008771193.1 | 3 | 277 | |||
| XM_008772972.1 | XP_008771194.1 | 3 | 279 | |||
| XM_008772973.1 | XP_008771195.1 | 3 | 203 | |||
| GenBank mRNA | BC158777.1 | - | 3 | 211 | 7133926 | |

Fam229b
Genomic Map