siRNA Details
| Gene Symbol: Smim11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131000.1 | NP_001124472.1 | 3 | 201 | ||
| XM_006248022.1 | XP_006248084.1 | 3 | 265 | |||
| GenBank mRNA | BC166910.1 | - | 3 | 202 | 6890105 | |
| BC166926.1 | - | 3 | 202 | 6919678 | ||
| FQ212222.1 | - | 3 | 231 | |||

Smim11
Genomic Map