siRNA Details
| Gene Symbol: Rup2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034950.2 | NP_001030122.2 | 2 | 194 | ||
| GenBank mRNA | AF198441.1 | - | 2 | 166 | ||
| AY327506.1 | - | 2 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rup2| Gene Symbol: Rup2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034950.2 | NP_001030122.2 | 2 | 194 | ||
| GenBank mRNA | AF198441.1 | - | 2 | 166 | ||
| AY327506.1 | - | 2 | 194 | |||