siRNA Details
| Gene Symbol: NCAPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281710.1 | NP_001268639.1 | 13 | 1677 | ||
| NM_001281711.1 | NP_001268640.1 | 13 | 1638 | |||
| NM_001281712.1 | NP_001268641.1 | 12 | 1457 | |||
| NM_015341.4 | NP_056156.2 | 13 | 1710 | |||
| XM_005263908.3 | XP_005263965.1 | 13 | 1650 | |||
| XM_006712388.3 | XP_006712451.1 | 13 | 1650 | |||
| GenBank mRNA | AK303725.1 | - | 12 | 1402 | ||
| AK308050.1 | - | 13 | 1617 | |||
| D38553.1 | - | 13 | 1640 | |||

NCAPH
Genomic Map