siRNA Details
| Gene Symbol: MIAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003491.3 | 3 | 418 | |||
| NR_033319.2 | 3 | 418 | ||||
| NR_033320.2 | 3 | 344 | ||||
| NR_033321.2 | 3 | 344 | ||||
| GenBank mRNA | AB263414.1 | - | 3 | 380 | ||
| AB263415.1 | - | 3 | 380 | |||
| AB263416.1 | - | 3 | 306 | |||
| AB263417.1 | - | 3 | 306 | |||
| AK098833.1 | - | 3 | 312 | |||
| DA213485.1 | - | 3 | 479 | |||

MIAT
Genomic Map