siRNA Details
| Gene Symbol: FRAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012083.2 | NP_036215.1 | 1 | 1219 | ||
| GenBank mRNA | AB045118.1 | - | 1 | 1219 | ||
| BC020165.1 | - | 1 | 1194 | 3629702 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FRAT2| Gene Symbol: FRAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012083.2 | NP_036215.1 | 1 | 1219 | ||
| GenBank mRNA | AB045118.1 | - | 1 | 1219 | ||
| BC020165.1 | - | 1 | 1194 | 3629702 | ||