siRNA Details
| Gene Symbol: KCNH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314030.1 | NP_001300959.1 | 5 | 864 | ||
| NM_012284.2 | NP_036416.1 | 5 | 868 | |||
| XM_011538085.2 | XP_011536387.1 | 5 | 1484 | |||
| XM_011538086.2 | XP_011536388.1 | 5 | 1480 | |||
| XM_017019096.1 | XP_016874585.1 | 4 | 1250 | |||
| XM_017019097.1 | XP_016874586.1 | 5 | 488 | |||
| GenBank mRNA | AB022696.1 | - | 5 | 643 | ||
| AB033108.1 | - | 5 | 868 | |||
| BC150602.1 | - | 5 | 749 | 9056972 | ||

KCNH3
Genomic Map