siRNA Details
| Gene Symbol: CBX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127321.1 | NP_001120793.1 | 3, 4 | 452 | ||
| NM_001127322.1 | NP_001120794.1 | 3, 4 | 650 | |||
| NM_012117.2 | NP_036249.1 | 3, 4 | 498 | |||
| GenBank mRNA | AK313506.1 | - | 3, 4 | 459 | ||
| BC006821.1 | - | 3, 4 | 400 | 3448801 | ||
| BP236173.1 | - | 3, 4 | 452 | |||
| BP336150.1 | - | 3, 4 | 498 | |||
| CR457418.1 | - | 2, 3 | 332 | |||
| EU794599.1 | - | 3, 4 | 467 | |||
| L07515.1 | - | 3, 4 | 467 | |||
| S62077.1 | - | 3, 4 | 466 | |||
| U26311.1 | - | 2, 3 | 330 | |||

CBX5
Genomic Map