siRNA Details
| Gene Symbol: CBX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127321.1 | NP_001120793.1 | 5 | 574 | ||
| NM_001127322.1 | NP_001120794.1 | 5 | 772 | |||
| NM_012117.2 | NP_036249.1 | 5 | 620 | |||
| GenBank mRNA | AK313506.1 | - | 5 | 581 | ||
| BC006821.1 | - | 5 | 522 | 3448801 | ||
| CR457418.1 | - | 4 | 454 | |||
| EU794599.1 | - | 5 | 589 | |||
| L07515.1 | - | 5 | 589 | |||
| S62077.1 | - | 5 | 588 | |||
| U26311.1 | - | 4 | 452 | |||

CBX5
Genomic Map