siRNA Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128208.1 | NP_001121680.1 | 3 | 342 | ||
| NM_015344.2 | NP_056159.2 | 3 | 342 | |||
| GenBank mRNA | AF063605.1 | - | 3 | 319 | ||
| AK075363.1 | - | 3 | 286 | |||
| AY277595.1 | - | 3 | 314 | |||
| AY358674.1 | - | 3 | 314 | |||
| BC000642.2 | - | 3 | 325 | 3344487 | ||
| CR541735.1 | - | 2 | 235 | |||
| CR541760.1 | - | 2 | 235 | |||
| DA258969.1 | - | 3 | 343 | |||

LEPROTL1
Genomic Map