siRNA Details
| Gene Symbol: FMR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024499.3 | 1 | 2329 | |||
| NR_024501.3 | 2 | 1947 | ||||
| NR_024502.3 | 3 | 1787 | ||||
| NR_024503.3 | 2 | 1683 | ||||
| GenBank mRNA | EU251065.1 | - | 1 | 2329 | ||
| EU251066.1 | - | 2 | 1947 | |||
| EU251067.1 | - | 3 | 1787 | |||
| EU251068.1 | - | 2 | 1683 | |||

FMR1-AS1
Genomic Map