siRNA Details
| Gene Symbol: TMEM131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015348.1 | NP_056163.1 | 34 | 4725 | ||
| XM_005263909.1 | XP_005263966.1 | 35 | 4811 | |||
| XM_005263910.1 | XP_005263967.1 | 35 | 4811 | |||
| XM_005263911.1 | XP_005263968.1 | 34 | 4694 | |||
| XM_005263912.3 | XP_005263969.1 | 33 | 4899 | |||
| XM_011510869.1 | XP_011509171.1 | 34 | 4625 | |||
| GenBank mRNA | AK025852.1 | - | 4 | 373 | ||
| AK098792.1 | - | 3 | 548 | |||
| D87446.1 | - | 33 | 4263 | |||

TMEM131
Genomic Map