siRNA Details
| Gene Symbol: TMEM131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015348.1 | NP_056163.1 | 27 | 3196 | ||
| XM_005263909.1 | XP_005263966.1 | 28 | 3282 | |||
| XM_005263910.1 | XP_005263967.1 | 28 | 3282 | |||
| XM_005263911.1 | XP_005263968.1 | 27 | 3165 | |||
| XM_005263912.3 | XP_005263969.1 | 26 | 3370 | |||
| XM_011510869.1 | XP_011509171.1 | 28 | 3282 | |||
| GenBank mRNA | D87446.1 | - | 26 | 2734 | ||

TMEM131
Genomic Map