siRNA Details
| Gene Symbol: KCTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015353.2 | NP_056168.1 | 3 | 535 | ||
| NR_110834.1 | 4 | 416 | ||||
| NR_110835.1 | 5 | 498 | ||||
| XM_011524575.2 | XP_011522877.1 | 3 | 497 | |||
| GenBank mRNA | AK307083.1 | - | 5 | 453 | ||
| BC033329.1 | - | 4 | 416 | |||
| BM906741.1 | - | 490 | ||||
| D79998.1 | - | 3 | 474 | |||

KCTD2
Genomic Map