siRNA Details
| Gene Symbol: NUP188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015354.2 | NP_056169.1 | 20 | 2025 | ||
| GenBank mRNA | BC040352.1 | - | 7 | 632 | ||
| BX537774.1 | - | 17 | 1669 | |||
| D79991.1 | - | 20 | 1987 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUP188| Gene Symbol: NUP188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015354.2 | NP_056169.1 | 20 | 2025 | ||
| GenBank mRNA | BC040352.1 | - | 7 | 632 | ||
| BX537774.1 | - | 17 | 1669 | |||
| D79991.1 | - | 20 | 1987 | |||