siRNA Details
| Gene Symbol: NUP188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015354.2 | NP_056169.1 | 22 | 2246 | ||
| GenBank mRNA | BC005407.1 | - | 3 | 187 | ||
| BC040352.1 | - | 9 | 853 | |||
| BX537774.1 | - | 19 | 1890 | |||
| D79991.1 | - | 22 | 2208 | |||

NUP188
Genomic Map