siRNA Details
| Gene Symbol: CDK20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039803.2 | NP_001034892.1 | 2, 3 | 527 | ||
| NM_001170639.1 | NP_001164110.1 | 2, 3 | 527 | |||
| NM_001170640.1 | NP_001164111.1 | 2, 3 | 527 | |||
| NM_012119.4 | NP_036251.2 | 2, 3 | 527 | |||
| NM_178432.3 | NP_848519.1 | 2, 3 | 566 | |||
| XM_017014561.1 | XP_016870050.1 | 2, 3 | 457 | |||
| XM_017014562.1 | XP_016870051.1 | 2, 3 | 457 | |||
| XM_017014563.1 | XP_016870052.1 | 2, 3 | 457 | |||
| XM_017014564.1 | XP_016870053.1 | 2, 3 | 457 | |||
| GenBank mRNA | AF035013.1 | - | 2, 3 | 430 | ||
| AF113130.1 | - | 2, 3 | 438 | |||
| AK075325.1 | - | 2, 3 | 319 | |||
| AK226135.1 | - | 1, 2 | 617 | |||
| AK291713.1 | - | 2, 3 | 448 | |||
| AK294193.1 | - | 2, 3 | 280 | |||
| AK296328.1 | - | 2, 3 | 278 | |||
| AK298993.1 | - | 2, 3 | 270 | |||
| AY904367.1 | - | 2, 3 | 193 | |||
| BC002655.2 | - | 2, 3 | 252 | 3605918 | ||
| BF966262.1 | - | 2, 3 | 505 | |||
| BI757318.1 | - | 2, 3 | 378 | |||
| BX647489.1 | - | 2, 3 | 456 | |||
| CB992644.1 | - | 2, 3 | 536 | |||
| KU178572.1 | - | 2, 3 | 193 | |||
| KU178573.1 | - | 2, 3 | 193 | |||
| KU178574.1 | - | 2, 3 | 193 | |||
| KU178575.1 | - | 2, 3 | 193 | |||
| KU178576.1 | - | 2, 3 | 193 | |||
| KU178577.1 | - | 2, 3 | 193 | |||

CDK20
Genomic Map